EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Association of Fecal Markers of Environmental Enteric Dysfunction with Zinc and Iron Status among Children at First Two Years of Life in Bangladesh.
PMID 29893201
Abstract
Environmental enteric dysfunction (EED) causes gut inflammation and increased intestinal permeability leading to deficiencies in micronutrients such as zinc and iron. Fecal markers such as myeloperoxidase (MPO), neopterin (NEO), and alpha-1-anti-trypsin (AAT) can predict EED. The aim of this study was to examine the association between fecal markers of EED with zinc and iron status among children at first 2 years of life. Malnutrition and Enteric Disease Study Bangladeshi birth cohort data were used to conduct this analysis. Multivariable analyses using generalized estimating equations were performed to test the association between individual fecal markers with zinc or iron status of the children. A total of 265 children were enrolled in the study (male:female = 1:1). Of the 627 stool samples collected (N = 222 children), 535, 511, and 577 were accompanied by zinc, ferritin, and soluble transferrin receptor values, respectively. Median (interquartile range [IQR]) values of AAT, MPO, and NEO were 0.33 (0.18-0.62) mg/g, 3,895.42 (1,563.76-8,432.82) ng/mL, and 890.81 (331.57-2,089.04) nmol/L, respectively. Overall, 60%, 71%, and 97% of samples were above the values considered normal in nontropical settings for AAT, MPO, and NEO, respectively. High AAT levels were significantly associated with low ferritin values after adjusting for age and gender (coefficient = -5.85; 95% confidence interval = -11.23 to -0.47; P value = 0.03). No such association was found between AAT and plasma zinc status. Myeloperoxidase and NEO were not associated with plasma zinc or iron status. The study results imply the importance of enteric protein loss in contributing to reduced ferritin levels at first 2 years of life.