EMAIL THIS PAGE TO A FRIEND
Macromolecular bioscience
Endothelialization and Anticoagulation Potential of Surface-Modified PET Intended for Vascular Applications.
PMID 29900675
Abstract
In vascular tissue engineering, great attention is paid to the immobilization of biomolecules onto synthetic grafts to increase bio- and hemocompatibility-two critical milestones in the field. The surface modification field of poly(ethylene terephthalate) (PET), a well-known vascular-graft material, is matured and oversaturated. Nevertheless, most developed methods are laborious multistep procedures generally accompanied by coating instability or toxicity issues. Herein, a straightforward surface modification procedure is presented engineered to simultaneously promote surface endothelialization and anticoagulation properties via the covalent immobilization of gelatin through a photoactivated azide derivative. A complete physicochemical characterization and biological study including cytotoxicity and endotoxin testing are performed. In addition, biocompatibility toward small (diameter ≤ 6 mm) and/or large caliber (diameter ≥ 6 mm) vessels is assessed by micro- and macrovascular endothelial cell assays. Superior bio- and hemocompatibility properties are seen for the gelatin-covalently modified PET surfaces compared to the conventional surface-modification procedures based on physisorption.