EMAIL THIS PAGE TO A FRIEND
Organic letters
Regioselective Synthesis of Fluorosulfonyl 1,2,3-Triazoles from Bromovinylsulfonyl Fluoride.
PMID 29906123
Abstract
A regioselective metal-free preparation of 4-fluorosulfonyl 1,2,3-triazoles from organic azides and a hitherto underexplored bromovinylsulfonyl fluoride building block is described. This reaction is very general and was extended to the synthesis of various sulfonates, sulfonamides, and sulfonic acid derivatives of triazoles and other azole heterocycles which would otherwise be difficult to access by existing methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
906190
1,2-Dibromoethane-1-sulfonyl fluoride
C2H3Br2FO2S