EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Mechanism of T-dependent cytotoxicity: role of papain-sensitive non class I MHC target molecules and expression of target antigen for cytotoxicity.
PMID 2994412