EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Effects of miR-200b-3p inhibition on the TRPC6 and BKCa channels of podocytes.
PMID 29958896
Abstract
Transient receptor potential canonical 6 (TRPC6) and large-conductance Ca2+-activated K+ channels (BKCa), two of the key ion channels for blood filtration function of podocytes, have been implicated in the pathogenesis of kidney diseases. Moreover, it has been reported that miR-200 b plays an important role in regulating the biological processes of podocytes. In this study, we aimed to examine whether there was a relationship between miR-200 b-3p and the two ion channels. It was suggested that miR-200 b-3p down-regulation inhibited the currents of TRPC6 and BKCa channels. It also showed that miR-200 b-3p inhibition reduced the levels of protein expression and mRNA transcription of TRPC6 and BKCa channels. Moreover, the down-regulation of miR-200 b-3p resulted in the decrease of the intracellular Ca2+ concentration. It was also suggested that the decrease of BKCa currents resulting from miR-200 b-3p inhibition could be regulated by TRPC6 channels. TRPC6 blockage also inhibited BKCa currents and reduced the level of BKCa expression. These results together suggested that miR-200 b-3p inhibition reduced the currents of TRPC6, which led to the decrease of intracellular Ca2+ concentration. The decrease of Ca2+ source required for BKCa activation may result in the inhibition of BKCa currents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O6754
1-Oleoyl-2-acetyl-sn-glycerol, ≥97% (TLC), oil
C23H42O5