EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Chirality transfer from organic ligands to silver nanostructures via chiral polarisation of the electric field.
PMID 29971311
Abstract
Water-soluble ligand protected optically active silver nanostructures were synthesised in a one-step reduction and capping process mediated by thiol-containing biomolecules. The synthesis was performed successfully with d- and l-cysteine and l-glutathione. The chiroptical properties of the obtained nanostructures were investigated by circular dichroism spectroscopy in the ultraviolet and visible wavelength range. They exhibit a g-value of up to 0.7%, which is about one order of magnitude larger compared to particles prepared by citrate reduction followed by a ligand exchange reaction. The structure and composition of the prepared materials were characterised by transmission electron microscopy, energy-dispersive X-ray and X-ray photoelectron spectroscopy. Although these structures do not have a chiral geometry, they show mirror image g-values when capped with d- and l-cysteine. This indicates that the underlying chirality transfer mechanism is based on an electric field polarisation process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30095
D-Cysteine, ≥99% (RT)
C3H7NO2S