EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Gonadotropin-releasing hormone receptors in human pituitary: ligand structural requirements, molecular size, and cationic effects.
PMID 2997260
Abstract
A specific, high affinity receptor for GnRH in human pituitaries obtained post mortem is described. The human pituitary GnRH receptor bound GnRH, a GnRH agonist [(D-Ala6,N alpha-MeLeu7,Pro9NEt)-GnRH], and a GnRH antagonist [Ac-D-Nal(2)1,D-alpha-Me-4-ClPhe2,D-3-Pal3,D-Arg6,D-Ala10 )-GnRH] with similar affinities (KdS of 4.81 nM, 0.32 nM, and 0.32 nM) to those of the rat pituitary (KdS of 4.71 nM, 0.31 nM, and 0.40 nM). A second GnRH antagonist [(D-pGlu1,D-Phe2,D-Trp3,6)-GnRH], however, was bound with a much lower affinity by the human receptor (Kd of 4.21 nM) than that of the rat (Kd of 0.09 nM). Monovalent and divalent cations affected [125I]GnRH agonist binding to rat and human pituitary receptors differently. In the presence of Mg2+ or Ca2+, binding to the human receptor was significantly lower than in the rat. At near physiological concentrations, Na+ and K+ (100 mM and 10 mM, respectively) inhibited [125I]GnRH agonist binding to the receptors to a similar extent in both species. At unphysiological concentrations (10 mM Na+ and 100 mM K+) these ions decreased binding to the human pituitary receptor to a greater extent than to the rat receptor. Using a ligand-immunoblotting technique, the human receptor or binding component of the receptor complex was found to be of greater molecular size (64,000 daltons) than that of the rat (60,000 daltons). The results show that the human and rat pituitary GnRH receptors have similar binding affinities for GnRH and certain GnRH analogs but differ in their binding of an antagonist, their sensitivity to cationic effects on GnRH agonist binding, and in the molecular size of the receptor GnRH-binding protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCP0180 [D-Lys6]-LH-RH
C59H84N18O13
SCP0181 [D-Lys6]-LH-RH Free Acid
C59H83N17O14
SCP0182 [D-Phe2,D-Ala6]-LH-RH
C59H79N15O13