EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Obesity-induced protein carbonylation in murine adipose tissue regulates the DNA-binding domain of nuclear zinc finger proteins.
PMID 30012885
Abstract
In obesity-linked insulin resistance, oxidative stress in adipocytes leads to lipid peroxidation and subsequent carbonylation of proteins by diffusible lipid electrophiles. Reduction in oxidative stress attenuates protein carbonylation and insulin resistance, suggesting that lipid modification of proteins may play a role in metabolic disease, but the mechanisms remain incompletely understood. Herein, we show that in vivo, diet-induced obesity in mice surprisingly results in preferential carbonylation of nuclear proteins by 4-hydroxy-trans-2,3-nonenal (4-HNE) or 4-hydroxy-trans-2,3-hexenal (4-HHE). Proteomic and structural analyses revealed that residues in or around the sites of zinc coordination of zinc finger proteins, such as those containing the C2H2 or MATRIN, RING, C3H1, or N4-type DNA-binding domains, are particularly susceptible to carbonylation by lipid aldehydes. These observations strongly suggest that carbonylation functionally disrupts protein secondary structure supported by metal coordination. Analysis of one such target, the nuclear protein estrogen-related receptor γ (ERR-γ), showed that ERR-γ is modified by 4-HHE in the obese state. In vitro carbonylation decreased the DNA-binding capacity of ERR-γ and correlated with the obesity-linked down-regulation of many key genes promoting mitochondrial bioenergetics. Taken together, these findings reveal a novel mechanistic connection between oxidative stress and metabolic dysfunction arising from carbonylation of nuclear zinc finger proteins, such as the transcriptional regulator ERR-γ.