EMAIL THIS PAGE TO A FRIEND
European journal of cancer (Oxford, England : 1990)
Indoleamine 2,3-dioxygenase 1 and programmed cell death-ligand 1 co-expression correlates with aggressive features in lung adenocarcinoma.
PMID 30014971
Abstract
Indoleamine 2,3-dioxygenase 1 (IDO1) is an immunosuppressive effector, and its expression is associated with prognosis in several cancer types. Here, we investigated the relationship between IDO1 expression in lung adenocarcinoma and patient prognosis and clinicopathological features, including programmed cell death-ligand 1 (PD-L1) expression. In this study, surgically resected primary lung adenocarcinoma specimens from 427 patients were evaluated for IDO1 and PD-L1 expression by immunohistochemistry, and lung adenocarcinoma cell lines were evaluated for IDO1 and PD-L1 protein expression by enzyme-linked immunosorbent assay and flow cytometry and for messenger RNA levels by real-time reverse-transcriptase polymerase chain reaction analysis. IDO1 was expressed in 260 patients (60.9%) at 1% cut-off and 63 patients (14.8%) at 50% cut-off. Tissues from 145 patients (34.0%) were positive for PD-L1 using the cut-off of 1%. Multivariate analysis showed that ≥1% IDO1 positivity was significantly associated with higher tumour grade, vascular invasion and PD-L1 expression. IDO1 and PD-L1 proteins were co-expressed in 123 patients (28.8%), and co-expressing tumours exhibited significantly more malignant traits than those positive for one or neither protein. In multivariate analysis, co-expression of IDO1 and PD-L1 was significantly associated with shorter disease-free survival and overall survival. Both proteins were upregulated in lung adenocarcinoma cell lines by treatment with interferon-γ and transforming growth factor-β. These results suggest that IDO1 and PD-L1 co-expression may define an aggressive form of lung adenocarcinoma.