EMAIL THIS PAGE TO A FRIEND
BMC anesthesiology
Mesenchymal stromal cells attenuate sevoflurane-induced apoptosis in human neuroglioma H4 cells.
PMID 30021512
Abstract
Inhalation of sevoflurane can induce neuronal apoptosis, cognitive impairment and abnormal behaviors. Bone marrow mesenchymal stem cells (MSCs) can secret neurotrophic factors and cytokines to protect from oxidative stress-related neuronal apoptosis. However, whether MSCs can protect from sevoflurane-induced neuronal apoptosis and the potential mechanisms are unclear. A non-contact co-culture of MSCs with human neuroglioma H4 cells (H4 cells) was built. H4 cells were co-cultured with MSCs or without MSCs (control) for 24 h. The co-cultured H4 cells were exposed to 4% sevoflurane for 6 h. The levels of caspase-3, reactive oxygen species (ROS), adenosine triphosphate (ATP), and the release of cytochrome C were determined by Western blot and fluorescence assay. Sevoflurane exposure significantly elevated the levels of cleaved caspase 3 and Bax in H4 cells. However, these phenomena were significantly offset by the co-culture with MSCs in H4 cells. Co-culture with MSCs before, but not after, sevoflurane exposure, significantly attenuated sevoflurane-induced ROS production in H4 cells. MSCs prevented sevoflurane-mediated release of cytochrome C from the mitochondria and production of ATP in H4 cells. Our study indicated that soluble factors secreted by MSCs attenuated the sevoflurane-induced oxidative stress and apoptosis of neuronal cells by preserving their mitochondrial function.