EMAIL THIS PAGE TO A FRIEND
Gastroenterologie clinique et biologique
[Hepatitis caused by suloctidil with antinuclear antibodies].
PMID 3002894
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9384
Suloctidil, analytical standard, for drug analysis
C20H35NOS