EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis and antiviral activity of (E)-5-(2-bromovinyl)uracil and (E)-5-(2-bromovinyl)uridine.
PMID 3005566
Abstract
(E)-5-(2-Bromovinyl)uracil (BVU) and (E)-5-(2-bromovinyl)uridine (BVRU) were synthesized starting from 5-formyluracil via (E)-5-(2-carboxyvinyl)uracil or starting from 5-iodouridine via (E)-5-(2-carbomethoxyvinyl)uridine and (E)-5-(2-carboxyvinyl)uridine, respectively. Depending on the choice of the cell system, BVU and BVRU exhibited a marked activity against herpes simplex virus type 1 (HSV-1) in vitro. Although BVU and BVRU were less potent than the reference compound (E)-5-(2-bromovinyl)-2'-deoxyuridine (BVDU), their antiviral activity spectrum was remarkably similar to that of BVDU. The latter findings suggest that BVU and BVRU are metabolically converted to BVDU or a phosphorylated product thereof. In vivo, BVU protected mice against a lethal disseminated HSV-1 infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9647
(E)-5-(2-Bromovinyl)-2′-deoxyuridine
C11H13BrN2O5
457442
(E)-5-(2-Bromovinyl)uracil, 97%
C6H5BrN2O2