EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Physicochemical Characterization, Antioxidant and Immunostimulatory Activities of Sulfated Polysaccharides Extracted from Ascophyllum nodosum.
PMID 30065217
Abstract
Polysaccharides from Ascophyllum nodosum (AnPS) were extracted and purified via an optimized protocol. The optimal extraction conditions were as follows: extraction time of 4.3 h, extraction temperature of 84 °C and ratio (v/w, mL/g) of extraction solvent (water) to raw material of 27. The resulting yield was 9.15 ± 0.23% of crude AnPS. Two fractions, named AnP1-1 and AnP2-1 with molecular weights of 165.92 KDa and 370.68 KDa, were separated from the crude AnPS by chromatography in DEAE Sepharose Fast Flow and Sephacryl S-300, respectively. AnP1-1 was composed of mannose, ribose, glucuronic acid, glucose and fucose, and AnP2-1 was composed of mannose, glucuronic acid, galactose and fucose. AnPS, AnP1-1 and AnP2-1 exhibited high scavenging activities against ABTS radical and superoxide radical, and showed protective effect on H₂O₂-induced oxidative injury in RAW264.7 cells. Furthermore, the immunostimulatory activities of AnP1-1 and AnP2-1 were evaluated by Caco-2 cells, the results showed both AnP1-1 and AnP2-1 could significantly promote the production of immune reactive molecules such as interleukin (IL)-8, IL-1β, interferon (IFN)-γ, and tumor necrosis factor (TNF)-α. Therefore, the results suggest that AnPS and its two fractions may be explored as a potential functional food supplement.