EMAIL THIS PAGE TO A FRIEND
Canadian journal of physiology and pharmacology
Studies of angiotensin I converting enzyme: effects of kinins, bacitracin, gamma-aminobutyric and epsilon-aminocaproic acids, and related compounds on substrate binding and catalysis in vitro.
PMID 3006897
Abstract
Bradykinin and 22 of its analogs were evaluated for their abilities to inhibit the hydrolysis of [3H]hippurylglycylglycine by purified porcine kidney angiotensin I converting enzyme. The mean inhibitory concentration (IC50) for bradykinin was 1.2 +/- 0.2 X 10(-6) M. Except for Ile-Ser-bradykinin and [Sar4]-bradykinin, none of the kinin analogs were more potent in this regard than bradykinin. Bacitracin, gamma-aminobutyric acid, epsilon-aminocaproic acid, and structurally related compounds were also tested. The IC50 value for bacitracin was 1.9 +/- 0.4 X 10(-4) M, gamma-aminobutyric acid, 83.4 +/- 7.2 mM, and for epsilon-aminocaproic acid, 7.0 +/- 1.4 mM. Compounds were also evaluated for their abilities to prevent 125I-labelled [Tyr1]-kallidin binding to angiotensin I converting enzyme inhibited by EDTA. The IC50 values for bradykinin, bacitracin, gamma-aminobutyric acid, and epsilon-aminocaproic acid were 1.6 +/- 0.3 X 10(-8) M, 2.6 +/- 0.9 X 10(-6) M, greater than 291 mM, and 13.2 +/- 3.9 mM, respectively.