EMAIL THIS PAGE TO A FRIEND
Plant science : an international journal of experimental plant biology
Combinatorial interaction of two adjacent cis-active promoter regions mediates the synergistic induction of Bt2 gene by sucrose and ABA in maize endosperm.
PMID 30080620
Abstract
The accumulation of starch in cereal endosperm is a key process that determines crop yield and quality. Research has reported that sucrose and abscisic acid (ABA) synergistically regulate the synthesis of crop starch. However, little is known about the molecular mechanisms behind this synergistic effect. In this study, the effect of sucrose and ABA on starch synthesis in maize endosperm was investigated. The starch content, the ADP-Glc pyrophosphorylase (AGPase) concentration, and the expression of AGPase-encoding genes were found to be enhanced slightly by sucrose or ABA alone, but were elevated significantly by the co-treatment of sucrose and ABA. Truncation analysis of the Bt2 promoter via transient expression in maize endosperm showed that the promoter region (-370/-186) is involved in sucrose response, and that an adjacent region (-186/-43) responds to ABA. The synergistic induction of sucrose and ABA on Bt2 promoter activity requires interaction with both of these regions. Interestingly, removal of the sucrose-responsive region (-370 to -186) abolishes ABA responsiveness in the Bt2 promoter, even in the presence of ABA-responsive region (-186 to -43). This study provides novel insights into the regulatory mechanisms that underlie the synergistic regulation of starch synthesis and grain filling from sucrose and ABA in cereal endosperm.