EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Peptide-directed liposomal delivery improves the therapeutic index of an immunomodulatory cytokine in controlling autoimmune arthritis.
PMID 30081142
Abstract
Rheumatoid arthritis (RA) is an autoimmune disease characterized by chronic inflammation of the synovial tissue of the joints. Inadequately controlled disease may cause severe joint damage and deformity. Currently, the anti-arthritic drugs are given systemically, and therefore, they are widely distributed to other organs that are not the intended therapeutic targets. Accordingly, using a particular dose/regimen of a drug to achieve an effective local concentration of the drug in arthritic joints may lead to expected adverse effects involving other organs. Thus, improved methods of drug delivery are needed for arthritis therapy. One attractive approach is the targeting of a systemically administered drug to the inflamed joints. We describe here a prototypic drug delivery system using a novel peptide ligand denoted as ART-1. We previously reported ART-1 (=ADK) as a peptide that preferentially homes to the inflamed joints of arthritic rats and binds to synovial endothelial cells. We tested the ART-1-coated liposomes encapsulating a fluorescent compound for binding to activated endothelial cells in vitro and homing to arthritic joints in vivo, compared to control liposomes lacking the ART-1 coating. Similar liposomes but encapsulating an immunomodulatory cytokine interleukin-27 (ART-1-IL-27 liposomes) were tested for their anti-arthritic activity compared with control liposomes. ART-1-displaying liposomes showed better binding to endothelial cells as well as in vivo homing to arthritic joints compared to control liposomes. Furthermore, ART-1-IL-27 liposomes, when intravenously injected to arthritic rats after the onset of arthritis, were more effective in suppressing disease progression than control-IL-27 liposomes lacking ART-1 or free IL-27 at an equivalent dose of IL-27. In addition, ART-1-directed liposomal IL-27 had a better safety profile than undirected liposomal IL-27 or free IL-27, thereby offering an improved therapeutic index for IL-27 therapy. These results provide a proof-of concept for the use of a novel joint-homing peptide for targeted delivery of drugs including biologics or small molecule compounds to arthritic joints with enhanced efficacy and reduced systemic exposure. This targeted therapy platform may be suitable for use in RA patients.