EMAIL THIS PAGE TO A FRIEND
Scientific reports
Evaluation of platelet indices as diagnostic biomarkers for colorectal cancer.
PMID 30087357
Abstract
Altered platelet indices, including platelet count (PC), mean platelet volume (MPV), platelet distribution width (PDW), and plateletcrit (PCT), have been found in various cancer types. This study aimed to evaluate the role of platelet indices as potential biomarkers for the diagnosis of colorectal cancer (CRC), and to assess the association between platelet indices and CRC clinicopathological characteristics. The study included 783 subjects with CRC, 463 subjects with colorectal adenomas (CA), and 689 control subjects from June 2015 to October 2017. All participants' clinicopathological characteristics were collected and analyzed. Here, we found that PC, MPV and PCT levels in CRC patients were significantly higher than those in CA patients and healthy participants (p < 0.001); however, PDW level in CRC patients was significantly higher than that in healthy participants while lower than that in CA patients. Receiver-operating characteristic (ROC) analysis indicated that combined detection of PCT and CEA appears to be a more effective marker to distinguish CRC patients from CA patients, with 70% sensitivity and 83% specificity. Among CRC patients, PC and PCT levels were associated with TNM stages and tumor size; MPV and PCT levels were associated with vascular invasion. Our findings suggest that altered PC, MPV and PCT levels might serve as potential biomarkers for the diagnosis and prognosis of CRC.