EMAIL THIS PAGE TO A FRIEND
Bioscience of microbiota, food and health
Stimulation of murine cell-mediated immunity by dietary administration of a cell preparation of Enterococcus faecalis strain KH-2 and its possible activity against tumour development in mice.
PMID 30094120
Abstract
It is well known that dietary lactic acid bacteria (LAB) stimulate cell-mediated immunity such as natural killer (NK) activity in mice. Here, we aimed to assay the immunomodulatory effects of a cell preparation of Enterococcus faecalis strain KH-2 (CPEF). We further evaluated the possibility of antitumour activity caused by CPEF administration, because NK cells actively participate in the prevention of tumour formation. NK cell activity and gene expression of IFN-γ and Perforin 1, which were induced most likely by a synergetic action of their cytotoxic activity, were higher in splenocytes of CPEF-administered mice than they were in control mice. Moreover, unlike those of control mice, the splenocytes of CPEF-administered mice had significantly higher CD28+CD69+/CD4+ and CD28+CD69+/CD8+ ratios that resulted in a survival rate with a tendency toward improvement after 47 days of CPEF administration (p=0.1) in Meth-A fibrosarcoma-bearing mice. In conclusion, we showed that CPEF might be effective in treating Meth-A fibrosarcoma in mice, as it helped increase their survival rate via stimulation of an immune response in splenocytes, which involved systemic cellular immunity processes such as cytotoxic activity, and active T cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4292
3,3′-Dioctadecyloxacarbocyanine perchlorate
C53H85ClN2O6