EMAIL THIS PAGE TO A FRIEND
Molecular and cellular endocrinology
Effect of oxidizing agents on rabbit mammary prolactin receptors.
PMID 3011564
Abstract
Treatment of rabbit mammary membrane-bound and solubilized prolactin receptors with the oxidizing agents N-chlorobenzene sulfonamide and N-bromosuccinimide resulted in total inactivation of the ability of the receptor to bind prolactin. A similar inactivation was obtained with 2-hydroxy-5-nitrobenzylbromide. The results suggest the possible importance of tryptophan in maintaining the activity of the prolactin receptor.
Related Materials