EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
Control of Peptide Aggregation and Fibrillation by Physical PEGylation.
PMID 30130095
Abstract
Peptide therapeutics have the potential to self-associate, leading to aggregation and fibrillation. Noncovalent PEGylation offers a strategy to improve their physical stability; an understanding of the behavior of the resulting polymer/peptide complexes is, however, required. In this study, we have performed a set of experiments with additional mechanistic insight provided by in silico simulations to characterize the molecular organization of these complexes. We used palmitoylated vasoactive intestinal peptide (VIP-palm) stabilized by methoxy-poly(ethylene glycol)5kDa-cholane (PEG-cholane) as our model system. Homogeneous supramolecular assemblies were found only when complexes of PEG-cholane/VIP-palm exceeded a molar ratio of 2:1; at and above this ratio, the simulations showed minimal exposure of VIP-palm to the solvent. Supramolecular assemblies formed, composed of, on average, 9-11 PEG-cholane/VIP-palm complexes with 2:1 stoichiometry. Our in silico results showed the structural content of the helical conformation in VIP-palm increases when it is complexed with the PEG-cholane molecule; this behavior becomes yet more pronounced when these complexes assemble into larger supramolecular assemblies. Our experimental results support this: the extent to which VIP-palm loses helical structure as a result of thermal denaturation was inversely related to the PEG-cholane:VIP-palm molar ratio. The addition of divalent buffer species and increasing the ionic strength of the solution both accelerate the formation of VIP-palm fibrils, which was partially and fully suppressed by 2 and >4 mol equivalents of PEG-cholane, respectively. We conclude that the relative freedom of the VIP-palm backbone to adopt nonhelical conformations is a key step in the aggregation pathway.