EMAIL THIS PAGE TO A FRIEND
No to shinkei = Brain and nerve
[Hypercoagulability and cerebral infarction].
PMID 3013261
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9384
Suloctidil, analytical standard, for drug analysis
C20H35NOS