Description
Molecular Formula
Add to Cart
577507-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
50494721 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
569220-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
569229-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 2.1 mm
50495521 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
504947-30 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 3 mm
504955-30 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm
504971-30 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
569222-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4 mm
569231-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
504955-40 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
504971-40 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
504947 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
569223-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
569232-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
504955 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
504971 Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
569224-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
569226-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
569234-U Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
505188 Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea
505161 Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
59576-U Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm, pkg of 2 ea
59575-U Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 3 mm
505137 Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
505129 Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm
55700-U21 Discovery® C18 Validation Pack, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
55700-U Discovery® C18 Validation Pack, 5 μm particle size, L × I.D. 5 cm × 4.6 mm