EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
The role of ozone and influence of band structure in WO3 photocatalysis and ozone integrated process for pharmaceutical wastewater treatment.
PMID 30144767
Abstract
Photocatalytic ozonation has great potential in wastewater treatment. However, the role of ozone and the contribution of photogenerated hole in this process have not been fully understood. Here three WO3 materials are synthesized and used as model catalysts in visible-light photocatalytic ozonation for the mineralization of pharmaceutical pollutants. A dual role of ozone in this process has been confirmed: (i) direct oxidation of the pollutant till formation of refractory intermediates, (ii) efficient trapping of photoelectron that cannot be captured by O2. The latter is crucial because it not only induces the O3--mediated pathway for hydroxyl radical (OH) formation but also separates the hole which has proven to be capable of oxidizing water into OH. Evidenced by photoluminescence results, the intrinsic charge separation ability of WO3 in photocatalytic ozonation is no more as important as that in photocatalysis with O2. Finally, this process is more applicable under acidic condition. This work contributes to a better understanding of the significance of ozone in WO3 photocatalytic ozonation and provides us an insight into the mechanism of photocatalytic ozonation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0675000 Cephalexin monohydrate, European Pharmacopoeia (EP) Reference Standard
C16H17N3O4S · H2O