EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Chitosan-magnetite nanocomposite as a sensing platform to bendiocarb determination.
PMID 30151686
Abstract
A novel platform for carbamate-based pesticide quantification using a chitosan/magnetic iron oxide (Chit-Fe3O4) nanocomposite as a glassy carbon electrode (GCE) modifier is shown for an analytical methodology for determination of bendiocarb (BND). The BND oxidation signal using GCE/Chit-Fe3O4 compared with bare GCE was catalyzed, showing a 37.5% of current increase with the peak potential towards less positive values, showing method's increased sensitivity and selectivity. Using square-wave voltammetry (SWV), calibration curves for BND determination were obtained (n = 3), and calculated detection and quantification limits values were 2.09 × 10-6 mol L-1 (466.99 ppb) and 6.97 × 10-6 mol L-1 (1555.91 ppb), respectively. The proposed electroanalytical methodology was successfully applied for BND quantification in natural raw waters without any sample pretreatment, proving that the GCE/Chit-Fe3O4 modified electrode showed great potential for BND determination in complex samples. ᅟ Graphical abstract.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45336
Bendiocarb, PESTANAL®, analytical standard
C11H13NO4