EMAIL THIS PAGE TO A FRIEND
Biomolecules
Assessment of the Quorum Sensing Inhibition Activity of a Non-Toxic Chitosan in an N-Acyl Homoserine Lactone (AHL)-Based Escherichia coli Biosensor.
PMID 30181497
Abstract
New approaches to deal with drug-resistant pathogenic bacteria are urgent. We studied the antibacterial effect of chitosans against an Escherichia coli quorum sensing biosensor reporter strain and selected a non-toxic chitosan to evaluate its quorum sensing (QS) inhibition activity and its effect on bacterial aggregation. To this end, chitosans of varying degree of acetylation (DA) (12 to 69%) and molecular weight (Mw) (29 to 288 kDa) were studied. Only chitosans of low DA (~12%) inhibited bacterial growth, regardless of their Mw. A chitosan with medium degree of polymerization (named MDP) DA30, with experimental DA 42% and Mw 115 kDa was selected for further QS inhibition and scanning electron microscopy (SEM) imaging studies. MDP DA30 chitosan exhibited QS inhibition activity in an inverse dose-dependent manner (≤12.5 µg/mL). SEM images revealed that this chitosan, when added at low concentration (≤30.6 µg/mL), induced substantial bacterial aggregation, whereas at high concentration (234.3 µg/mL), it did not. Aggregation explains the QS inhibition activity as the consequence of retardation of the diffusion of N-acylated homoserine lactones (AHLs).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K3007
N-(β-Ketocaproyl)-L-homoserine lactone, ≥98%
C10H15NO4