EMAIL THIS PAGE TO A FRIEND
Neuro endocrinology letters
Effects of alprostadil combined with calcium dobesilate in patients with diabetic peripheral neuropathy.
PMID 30183208
Abstract
To observe the clinical curative effects of alprostadil combined with calcium dobesilate in type 2 diabetes patients with peripheral neuropathy. We randomly divided 120 type 2 diabetes patients with diabetic peripheral neuropathy into two groups. The treatment group was prescribed alprostadil (10 μg, once daily) and oral calcium dobesilate (0.5 g, 3 times daily), and the control group was prescribed alprostadil (10 μg, once daily) for a total treatment duration of 2 weeks. The Michigan Diabetic Neuropathy Score (MDNS) and the Michigan Neuropathy Screening Instrument (MNSI) were used to evaluate differences between the two groups before and after treatment. Following 2 weeks of treatment, the total effective rate in the treatment group was significantly better than that of the control group (p<0.05) and the MDNS and MNSI scores in the treatment group were significantly lower than those in the control group (p<0.05 or p<0.01). Combined alprostadil and calcium dobesilate treatment for type 2 diabetic peripheral neuropathy showed good clinical efficacy and an improved curative effect than single alprostadil treatment.