EMAIL THIS PAGE TO A FRIEND
Photochemistry and photobiology
Free radical production by chlorpromazine sulfoxide, an ESR spin-trapping and flash photolysis study.
PMID 3018808
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000507 Chlorpromazine impurity A, European Pharmacopoeia (EP) Reference Standard
C17H19ClN2OS