EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Directing the nanoparticle formation by the combination with small molecular assembly and polymeric assembly for topical suppression of ocular inflammation.
PMID 30213683
Abstract
In this paper, we presented a simple yet versatile strategy to generate a high drug payload nanoparticles by the combination with small molecular assembly and polymeric assembly for topical suppression of ocular inflammation. Upon physical mixing of the succinated triamcinolone acetonide (TA-SA) supramolecular hydrogel with the poly (ethylene glycol)-poly (ɛ-caprolactone)-poly (ethylene glycol) (PECE) aqueous solution at 37 °C, TA-SA/PECE nanoparticles formed spontaneously and characterized thoroughly by transmission electron microscopy (TEM), X-ray diffraction (XRD) and differential scanning calorimetry (DSC). The formed TA-SA/PECE nanoparticles displayed a comparable in vitro anti-inflammatory efficacy to that of native triamcinolone acetonide (TA), through a significant downregulation of various proinflammatory cytokines levels (e.g., NO, TNF-α) in a lipopolysaccharide (LPS) actived RAW264.7 macrophage. Meanwhile, the enhanced transcorneal drug permeability of TA-SA/PECE nanoparticles over that of TA suspension was clearly observed in an isolated rabbit cornea. Intraocular biocompatibility test demonstrated that TA-SA/PECE nanoparticles presented good biocompatibility after topical instillation during entire study period. More importantly, the TA-SA/PECE nanoparticles displayed superior therapeutic efficacy over that of the TA suspension in the endotoxin-induced uveitis (EIU) rabbit model via decreasing neutrophil infiltration in anterior chamber. Overall, the proposed TA-SA/PECE nanoparticles might be a promising candidate for uveitis therapy.