EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
The silencing of ApoC3 suppresses oxidative stress and inflammatory responses in placenta cells from mice with preeclampsia via inhibition of the NF-κB signaling pathway.
PMID 30257353
Abstract
Preeclampsia is one of the three primary causes of maternal morbidity and mortality worldwide. This study evaluated ApoC3 in placenta cells of mice with preeclampsia to explore its therapeutic role in preeclampsia and assess its function on oxidative stress and inflammatory responses involving the NF-κB signaling pathway. A mouse model of preeclampsia was successfully established. APOC3-siRNA with the best silencing effect was screened out. The expression levels of ApoC3, p65, and IkBα were evaluated. The effect of ApoC3 silencing on metabolic activity and apoptosis was measured. The level of high-sensitivity C-reactive protein (hs-CPR), interleukin 6 (IL-6) and tumor necrosis factor alpha (TNF-α), the activity of matrix metalloproteinase (MMP)-2 and MMP-9, and the expression of malondialdehyde (MDA), 8-isoprostane and oxidized low-density lipoprotein (ox-LDL) were determined. ApoC3-siRNA-3 was the most effective siRNA. The mRNA expression of ApoC3 was scarcely observed, while the expression of p65 decreased and the expression of p-IkBα increased in the ApoC3-siRNA group. Compared with those in the model and empty vector groups, the cell apoptosis rate and the activities of invasion-related factors MMP-2 and MMP-9 increased, while the levels of hs-CPR, IL-6, TNF-α, MDA, 8-isoprostane, and ox-LDL decreased in the ApoC3-siRNA group. Silencing ApoC3 could suppress the NF-κB signaling pathway, thereby exercising a protective effect on cell injury induced by oxidative stress and reducing inflammatory responses.