Description
Molecular Formula
Add to Cart
57264-U IonSense® C18 SPE-it Tips, pkg of 96 ea
57249-U
IonSense® PDMS/DVB SPE-it Tips, pkg of 96 ea
57557-U SPME FFA PTFE Sealing Cups, size 23ga Beveled (No line)
57562-U SPME FFA PTFE Sealing Cups, 23ga BLUNT, size 23ga Blunt (1 Line)
57561-U SPME FFA PTFE Sealing Cups, 24ga BLUNT, size 24ga blunt (2 Lines)
57592-U
SPME FFA Storage Device, for use with SPME FFA
57355-U SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm, metal alloy fiber, for use with manual holder
57354-U SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm, for use with autosampler
FFA57354-U SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm, for use with multi fiber exchanger
57306 SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
57307 SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
57285-U SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
FFA57285-U SPME fiber assortment kit 1, for use with multi fiber exchanger, needle size 23 ga
57320-U SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
57321-U SPME fiber assortment kit 2, for use with autosampler, size 24 ga
57286-U SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
FFA57286-U SPME fiber assortment kit 2, needle size 23 ga, for use with multi fiber exchanger
57323-U SPME fiber assortment kit 3
57324-U SPME fiber assortment kit 4, for use with manual holder, size 24 ga
57325-U SPME fiber assortment kit 4, for use with autosampler, size 24 ga
57287-U SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
FFA57287-U SPME fiber assortment kit 4, for use with multi fiber exchanger, needle size 23 ga
57362-U SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
FFA57362-U SPME fiber assortment kit 5, needle size 23 ga, for use with multi fiber exchanger
57234-U
SPME LC Tips, C18, tray of 96 ea
57248-U
SPME LC Tips, PDMS/DVB, pkg of 96 ea
57357-U SPME Sampling Stand, for use with 15 mL vials
57333-U SPME Sampling Stand, for use with 4 mL vials
57550-U SPME StableFlex fiber assortment kit, for use with manual holder
57551-U SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler