EMAIL THIS PAGE TO A FRIEND
Cell
SnapShot: RNA Structure Probing Technologies.
PMID 30290145
Abstract
Chemical probing coupled to high-throughput sequencing offers a flexible approach to uncover many aspects of RNA structure relevant to its cellular function. With a wide variety of chemical probes available that each report on different features of RNA molecules, a broad toolkit exists for investigating in vivo and in vitro RNA structure and interactions with other molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
908401
1-Methyl-7-nitroisatoic anhydride
C9H6N2O5
913839
2-Methylnicotinic acid imidazolide
C10H9N3O