EMAIL THIS PAGE TO A FRIEND
Archives internationales de pharmacodynamie et de therapie
Effect of triflusal and other salicylic acid derivatives on cyclic AMP levels in rat platelets.
PMID 3030206
Abstract
The effect of triflusal, acetylsalicylic acid (ASA), and of their principal metabolites 2-hydroxy-4-trifluoromethylbenzoic acid (HTB) and salicylic acid (SA), alone or combined with dypiridamole (DIP) and/or PGE1 on cyclic AMP levels in washed rat platelets (37 degrees C, 4 min), has been determined. DIP at 0.1 mM increased cyclic AMP levels by 25%. The effect of triflusal and HTB was significant at therapeutic concentrations of triflusal (1 mM: 36% increase) and HTB (0.5 mM: 37% increase). The effect of HTB was always greater than that of triflusal. ASA, at 1 mM and 5 mM, alone or combined with PGE1 was without effect. When 1 mM triflusal was combined with 0.1 mM DIP an increased effect was obtained (95%). ASA, at the highest concentration tested (5 mM), did not modify the DIP-induced increase of cyclic AMP levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2005010 Triflusal impurity B, European Pharmacopoeia (EP) Reference Standard
C8H5F3O3