EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Determination of kininase I and kininase II activities in human urine by high-performance liquid chromatography.
PMID 3033004
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6875
Hippuryl-L-phenylalanine
C18H18N2O4