EMAIL THIS PAGE TO A FRIEND
International journal of radiation biology and related studies in physics, chemistry, and medicine
Isolation of 2-deoxy-D-erythro-pentonic acid from an alkali-labile site in gamma-irradiated DNA.
PMID 303634
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14699
2-Deoxy-D-ribonic acid lithium salt, ≥95.0% (TLC)
C5H10O5 · xLi+