Description
Molecular Formula
Add to Cart
57264-U IonSense® C18 SPE-it Tips, pkg of 96 ea
57249-U
IonSense® PDMS/DVB SPE-it Tips, pkg of 96 ea
57557-U SPME FFA PTFE Sealing Cups, size 23ga Beveled (No line)
57562-U SPME FFA PTFE Sealing Cups, 23ga BLUNT, size 23ga Blunt (1 Line)
57561-U SPME FFA PTFE Sealing Cups, 24ga BLUNT, size 24ga blunt (2 Lines)
57592-U
SPME FFA Storage Device, for use with SPME FFA
57318 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, for use with manual holder, needle size 24 ga
57344-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 23 ga, for use with manual holder
57319 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 24 ga, for use with autosampler
57343-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 23 ga, for use with autosampler
57334-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, for use with manual holder, StableFlex fiber
57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, StableFlex, for use with autosampler
57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, StableFlex, for use with autosampler
57906-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, for use with autosampler, needle size 23 ga, metal alloy fiber
FFA57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, Stableflex, for use with multi fiber exchanger
FFA57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, for use with multi fiber exchanger, needle size 24 ga, StableFlex
57294-U SPME fiber assembly polyacrylate (PA), df 85 μm, for use with autosampler, needle size 23 ga
FFA57294-U SPME fiber assembly polyacrylate (PA), df 85 μm, for use with multi fiber exchanger, needle size 23 ga
57302 SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm, needle size 24 ga, for use with manual holder
57303 SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm, needle size 24 ga, for use with autosampler
57308 SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm, needle size 24 ga, for use with manual holder
57309
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm, needle size 24 ga, for use with autosampler
FFA57289-U SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm, needle size 23 ga, for use with multi fiber exchanger
FFA57291-U SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm, needle size 23 ga, for use with multi fiber exchanger
FFA57302 SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm, for use with multi fiber exchanger, needle size 24 ga
FFA57309 SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm, needle size 24 ga, for use with multi fiber exchanger
57355-U SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm, metal alloy fiber, for use with manual holder
57354-U SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm, for use with autosampler
FFA57354-U SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm, for use with multi fiber exchanger
57306 SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga