Description
Molecular Formula
Add to Cart
57264-U IonSense® C18 SPE-it Tips, pkg of 96 ea
57249-U
IonSense® PDMS/DVB SPE-it Tips, pkg of 96 ea
57557-U SPME FFA PTFE Sealing Cups, size 23ga Beveled (No line)
57562-U SPME FFA PTFE Sealing Cups, 23ga BLUNT, size 23ga Blunt (1 Line)
57561-U SPME FFA PTFE Sealing Cups, 24ga BLUNT, size 24ga blunt (2 Lines)
57592-U
SPME FFA Storage Device, for use with SPME FFA
57318 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, for use with manual holder, needle size 24 ga
57344-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 23 ga, for use with manual holder
57319 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 24 ga, for use with autosampler
57343-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 23 ga, for use with autosampler
57334-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, for use with manual holder, StableFlex fiber
57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, StableFlex, for use with autosampler
57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, StableFlex, for use with autosampler
57906-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, for use with autosampler, needle size 23 ga, metal alloy fiber
FFA57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, Stableflex, for use with multi fiber exchanger
FFA57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, for use with multi fiber exchanger, needle size 24 ga, StableFlex
57328-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
57348-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with manual holder/autosampler
57329-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
57299-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder/autosampler, fiber L 2 cm
57298-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
57912-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
57914-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
FFA57298-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, for use with multi fiber exchanger, Stableflex
FFA57329-U SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with multi fiber exchanger
57304 SPME fiber assembly polyacrylate (PA), df 85 μm, for use with manual holder, needle size 24 ga
57305 SPME fiber assembly polyacrylate (PA), df 85 μm, for use with autosampler, needle size 24 ga
57294-U SPME fiber assembly polyacrylate (PA), df 85 μm, for use with autosampler, needle size 23 ga
FFA57294-U SPME fiber assembly polyacrylate (PA), df 85 μm, for use with multi fiber exchanger, needle size 23 ga
57302 SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm, needle size 24 ga, for use with manual holder