EMAIL THIS PAGE TO A FRIEND
Prostaglandins, leukotrienes, and medicine
Altered arachidonic acid metabolism in granulocytes of polytraumatized patients.
PMID 3039535
Abstract
The generation and metabolism of leukotrienes (LTs) from polymorphonuclear granulocytes of four polytraumatic patients on stimulation with the Ca-Ionophore A 23187 were studied by high performance liquid chromatography. In contrast to healthy donors the concentration of LTB4 within the supernatant of the stimulated granulocytes from these patients is reduced. The ratio of LTB4 versus the combined amounts of the biologically inactive 6-trans and 12-epi-6-trans-isomers is significantly decreased. In one patient who suffered from an Adult Respiratory Distress Syndrome (ARDS) a pronounced enhancement of LTC4 synthesis was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6046
20-Carboxy-leukotriene B4, ~1 mg/mL in ethanol, ≥90% (HPLC)
C20H30O6