EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Purification and properties of an extracellular collagenolytic protease produced by the human oral bacterium Bacillus cereus (strain Soc 67).
PMID 3040751
Abstract
The major collagenolytic proteinase present in the culture filtrate of Bacillus cereus (strain Soc 67, isolated from the human oral cavity) has been purified to homogeneity by a procedure that comprised concentration of ultrafiltered growth medium on a Millipore PTTK00005 membrane, precipitation with ammonium sulfate, gel permeation chromatography, chromatofocusing, fast protein liquid chromatography on an anion-exchange column, and finally fast protein liquid chromatography on a gel column. The enzyme hydrolyzed, with decreasing rates, phenylazobenzyloxy-carbonyl-L-Pro-L-Leu Gly-L-Pro-D-Arg (PZ-PLGPA), furylacrylolyl-L-Leu-Gly-L-Pro-L-Ala, and furylacryloyl-L-Phe-Gly-Gly, while furylacryloyl-Gly-L-Leu-NH2 was not hydrolyzed. The enzyme degraded soluble and insoluble collagens, Azocoll and gelatin. Bradykinin was hydrolyzed at a high rate at the Phe-Ser bond. The enzyme was sensitive to pyrophosphate, L-cysteine, and L-histidine and could be totally inactivated in the presence of metal chelators. The enzyme contains 1 mol of Zn/mol and the hydrolysis of PZ-PLGPA is slightly increased by Ca2+. The enzyme is readily inhibited by heavy metal cations, but Cu2+ and Ni2+ affected the catalysis in opposite ways: increasing levels of Cu2+ decreased the affinity of the enzyme for PZ-PLGPA, whereas Ni2+ had no effect. The effect of Cu2+ also depended on the pH and type of buffer used. Detailed chemical modification experiments suggested that the active site of the enzyme contains at least 1 tyrosyl and 1 lysyl residue, and 1 carboxyl group. The enzyme was not sensitive to sulfhydryl reagents and thiols did not activate the enzyme. The modification studies were unable to reveal active histidyl residues. The ability of the enzyme to hydrolyze PZ-PLGPA, furylacryloyl-L-Leu-Gly-L-Pro-L-Ala, furylacryloyl-L-Phe-Gly-Gly, and various collagenous materials, its inactivity toward furylacryloyl-Gly-L-Leu-NH2, and the results from the chemical modification studies suggest that the B. cereus (Soc 67) collagenolytic enzyme can be regarded as a true collagenase which resembles the Clostridium histolyticum collagenase(s).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1889
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat hepatocytes tested, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C5894
Collagenase from Clostridium histolyticum, sterile; sterile-filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C1639
Collagenase from Clostridium histolyticum, sterile-filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C1764
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat epididymal adipocytes tested, Type II-S, 0.5-5.0 FALGPA units/mg solid
C9722
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
C8051
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
C0773
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, 1,000-3,000 CDU/mg solid (CDU = collagen digestion units)
C9697
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), Suitable for digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
C2014
Collagenase from Clostridium histolyticum, sterile-filtered, suitable for release of physiologically active rat pancreatic islets, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
C0255
Collagenase from Clostridium histolyticum, purified by chromatography, ≥0.25 FALGPA units/mg solid, ≥250 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
C2399
Collagenase from Clostridium histolyticum, sterile-filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, 1,000-3,000 CDU/mg solid
C7926
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
C2799
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, 1000-3000 CDU/mg solid (CDU = collagen digestion units), ≥4 FALGPA units/mg solid, high purity
C4785
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat pancreatic islets tested, Type XI-S, 2-5 FALGPA units/mg solid, >1200 CDU/mg solid
C8176
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
F5135
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala
C23H32N4O7