EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Novel compound for identifying Escherichia coli.
PMID 3046494
Abstract
A new chromogenic compound, 5-bromo-4-chloro-3-indoxyl-beta-D-glucuronide, was found to be useful for the rapid, specific, differential identification of Escherichia coli in the sanitary analysis of shellfish and wastewater. Of 1,025 presumptively positive colonies (blue) and 583 presumptively negative colonies (nonblue), only 1% false-negative and 5% false-positive results were found.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4527
5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside, ≥97%
C14H15BrClNO6