EMAIL THIS PAGE TO A FRIEND
Blood
Identification of a second transforming gene, rasn, in a human multiple myeloma line with a rearranged c-myc allele.
PMID 3048435
Abstract
Multiple myeloma is a disease characterized by a long, slowly progressive phase and a final, more aggressive one. Little is known about the mechanism of transformation of myeloma cells, although the clinical characteristics of the disease suggest a multi-step process. Recently, a myeloma cell line, NCI-H929, was isolated from a patient with aggressive preterminal disease and found to have a rearranged myc allele. This myeloma cell line has been further characterized in a focus formation assay to determine whether its unusual growth characteristics were associated with a second activated transforming gene. We now report that the NCI-H929 myeloma cell line has an activated rasn allele in addition to a rearranged myc allele. This is the first identification of an activated transforming gene in a multiple myeloma cell line; furthermore, the characterization of two independently activated oncogenes in this B cell malignancy has implications for both the pathogenesis and evolution of the disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
NCI-H929, 95050415