EMAIL THIS PAGE TO A FRIEND
Annual review of biochemistry
Molecular organization and function of the complement system.
PMID 3052276
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5938 Complement factor I from human plasma, >90% (SDS-PAGE)