EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Cholecystokinin receptor antagonists.
PMID 3054325
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CBR00275
5-Amino-5-oxopentanoic acid, AldrichCPR
C5H9NO3