EMAIL THIS PAGE TO A FRIEND
Acta odontologica Scandinavica
On the role of calcium fluoride in the cariostatic mechanism of fluoride.
PMID 3063053
Abstract
The literature concerning the formation and stability of CaF2 in the oral environment is reviewed. In early work the CaF2 formed during topical application with fluoride was assumed to be beneficial. It was suggested that it could protect the enamel surface directly or provide free fluoride ions for subsequent incorporation into the hydroxyapatite lattice. However, McCann claimed, in 1968, that CaF2 is soluble in saliva (12-15 mg/l), that it would be rapidly lost in the oral cavity, and that the clinical effect of fluoride was related to formation of firmly bound fluoride only. In this period many authors reported total loss of CaF2 during 24 h after a topical application of fluoride. It has now been shown in several laboratories that calcium fluoride is stable in saliva at neutral pH owing to surface adsorption of HPO2-4 to the crystal surface and formation of a solubility-limiting phase. Extended exposure of saliva can cause formation of a fluorapatite layer on the CaF2 crystals, restricting their dissolution further. Low pH (pH less than 5) causes loss of the solubility-limiting adsorbed HPO2-4 and a slow dissolution of CaF2. The CaF2 crystals may thus serve as pH-controlled reservoirs of fluoride ions on the enamel or in plaque and release fluoride during caries challenges. It is suggested that calcium fluoride is an essential phase explaining important aspects of the mechanism of topically applied fluoride, contrary to what was assumed in the past.