EMAIL THIS PAGE TO A FRIEND
Journal de radiologie
[Iohexol in studies of the spinal canal. Multicenter study].
PMID 3070021
Abstract
The author presents the results of a multicentric study of Iohexol in investigation of the spinal canal undertaken at the request of Winthrop Laboratories in 32 Radiological departments. The study involved 329 adults of both sexes. It confirmed the excellent quality of results obtained with this preparation in the literature and its excellent tolerance. On the basis of these results Iohexol may be considered to be definitely one of the best currently available preparations for investigation of the spinal canal.