EMAIL THIS PAGE TO A FRIEND
Clinical therapeutics
A comprehensive review of the clinical pharmacology and pharmacokinetics of cefaclor.
PMID 3072086
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72579
Cefaclor, analytical standard
C15H14ClN3O4S