EMAIL THIS PAGE TO A FRIEND
Clinical therapeutics
Cefaclor in the treatment of lower respiratory tract infections: a review.
PMID 3072087
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72579
Cefaclor, analytical standard
C15H14ClN3O4S