EMAIL THIS PAGE TO A FRIEND
Trends in biochemical sciences
Thus started ganglioside research.
PMID 3075367
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2250 Gangliosides Purified from bovine brain, Type II