EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Gel chromatography of pyridoxalated and glutaraldehyde-treated human haemoglobin on Superose 12.
PMID 3086347
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9514 Superose® 12 Prep Grade