EMAIL THIS PAGE TO A FRIEND
Urology
Effect of renacidin on suture material.
PMID 3097902
Abstract
Renacidin solution is used sometimes to irrigate renal units with retained calculi after definitive stone surgery. The effects of this solution on the integrity of suture material utilized to close kidneys have not been determined previously. In an attempt to define the best suture material to use when postoperative renacidin irrigation is contemplated, we studied the effects of this solution on a variety of sutures commonly used in genitourinary surgery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1928
Fenaminosulf, ≥98%
C8H10N3NaO3S
45482
Fenaminosulf, PESTANAL®, analytical standard
C8H10N3NaO3S