EMAIL THIS PAGE TO A FRIEND
British medical journal (Clinical research ed.)
Renal failure and the histopathological features of myeloma kidney reversed by intensive chemotherapy and peritoneal dialysis.
PMID 3101900
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3046 GDF-15 human